Anti-RSV Human Recombinant Monoclonal Antibody [clone: RV11]
New Product
This antibody has been tested to bind to fusion (F) glycoprotein of human Respiratory Syncytial Virus (RSV) subtype A1 strain (Cat. No. 77761-674) or B1 strain (Cat. No. 77761-676). An example of comparison of IgM and IgG using size exclusion chromatography is shown.
Type: Primary
Antigen: RSV
Clonality: Recombinant Monoclonal
Clone: RV11
Conjugation: Unconjugated
Epitope:
Host: Human
Isotype: IgM kappa
Reactivity: Human
- Catalog No:
- 77761-684
- 77761-686
- Antigen Symbol:
- RSV
- RSV
- Antigen Name:
- Respiratory syncytial virus
- Respiratory syncytial virus
- Antigen Synonyms:
- Fusion (F) glycoprotein
- Fusion (F) glycoprotein
- Antibody Type:
- Primary
- Primary
- Clonality:
- Recombinant Monoclonal
- Recombinant Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- RV11
- RV11
- Reactivity:
- Human
- Human
- Host:
- Human
- Human
- Isotype:
- IgM kappa
- IgM kappa
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Neutralisation:
- Yes
- Yes
- Concentration:
- 1 mg/ml by Refractometry
- 1 mg/ml by Refractometry
- Storage Buffer:
- PBS, pH 7.2
- PBS, pH 7.2
- Storage Temperature:
- Store at 4 °C, −20 °C
- Store at 4 °C, −20 °C
- Shipping Temperature:
- Ice
- Ice
- SizePackaging
- 77761-684L46X29-100UG100 µgVialEach(1/Itm/Bx)$997.02New Product
Specifications
Antigen SymbolRSVAntigen NameRespiratory syncytial virusEnvironmentally PreferableAntigen SynonymsFusion (F) glycoproteinAntibody TypePrimaryClonalityRecombinant MonoclonalConjugationUnconjugatedCloneRV11ReactivityHumanHostHumanIsotypeIgM kappaWestern BlotYesELISAYesImmunoChemistryYesImmunoPrecipitationYesImmunoFluorescenceYesFlow CytometryYesNeutralisationYesConcentration1 mg/ml by RefractometryStorage BufferPBS, pH 7.2Storage TemperatureStore at 4 °C, −20 °CShipping TemperatureIceNew Product - New Product
Specifications
Antigen SymbolRSVAntigen NameRespiratory syncytial virusEnvironmentally PreferableAntigen SynonymsFusion (F) glycoproteinAntibody TypePrimaryClonalityRecombinant MonoclonalConjugationUnconjugatedCloneRV11ReactivityHumanHostHumanIsotypeIgM kappaWestern BlotYesELISAYesImmunoChemistryYesImmunoPrecipitationYesImmunoFluorescenceYesFlow CytometryYesNeutralisationYesConcentration1 mg/ml by RefractometryStorage BufferPBS, pH 7.2Storage TemperatureStore at 4 °C, −20 °CShipping TemperatureIceNew Product