Anti-POLK Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
New Product
Type:
Antigen: POLK
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77842-668
- 77842-669
- Antigen Symbol:
- POLK
- POLK
- Antigen Name:
- DNA polymerase kappa protein (227-337AA)
- DNA polymerase kappa protein (227-337AA)
- Antigen Synonyms:
- DinB homolog 1 (E. coli)|DINB protein|DINB1|DINP|DNA damage inducible protein b|DNA polymerase kappa|Polk|POLK_HUMAN|POLQ|Polymerase (DNA directed) kappa|polymerase, DNA, kappa
- DinB homolog 1 (E. coli)|DINB protein|DINB1|DINP|DNA damage inducible protein b|DNA polymerase kappa|Polk|POLK_HUMAN|POLQ|Polymerase (DNA directed) kappa|polymerase, DNA, kappa
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UBT6
- Q9UBT6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
- SizePackaging
- 77842-668A70528-100UG100 µgVialEach(1/Itm/Bx)$330.00New Product
Specifications
Antigen SymbolPOLKAntigen NameDNA polymerase kappa protein (227-337AA)Environmentally PreferableAntigen SynonymsDinB homolog 1 (E. coli)|DINB protein|DINB1|DINP|DNA damage inducible protein b|DNA polymerase kappa|Polk|POLK_HUMAN|POLQ|Polymerase (DNA directed) kappa|polymerase, DNA, kappaClonalityRecombinant PolyclonalConjugationHRP (Horseradish Peroxidase)ReactivityHumanHostRabbitGene IDQ9UBT6IsotypeIgGELISAYesFormLiquidPurification>95%, Protein G purifiedStorage Buffer0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4Storage TemperatureUpon receipt, store at −20 or −80 °C. Avoid repeated freeze.Shipping TemperatureShipped on blue iceNew Product - New Product
Specifications
Antigen SymbolPOLKAntigen NameDNA polymerase kappa protein (227-337AA)Environmentally PreferableAntigen SynonymsDinB homolog 1 (E. coli)|DINB protein|DINB1|DINP|DNA damage inducible protein b|DNA polymerase kappa|Polk|POLK_HUMAN|POLQ|Polymerase (DNA directed) kappa|polymerase, DNA, kappaClonalityRecombinant PolyclonalConjugationHRP (Horseradish Peroxidase)ReactivityHumanHostRabbitGene IDQ9UBT6IsotypeIgGELISAYesFormLiquidPurification>95%, Protein G purifiedStorage Buffer0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4Storage TemperatureUpon receipt, store at −20 or −80 °C. Avoid repeated freeze.Shipping TemperatureShipped on blue iceNew Product