Boron, Plasma Standard Solution, Specpure®, B 1000 µg/ml, Thermo Fisher Scientific

Supplier: Thermo Fisher Scientific
Specpure®
39147-AD 39147-AE 039147-AP 39147-AP 039147-AD 039147-AE
AA39147-APEA 164.34 USD
CAAA39147-AP AA39147-AP CAAA39147-AD CAAA39147-AE AA39147-AD AA39147-AE
Boron, Plasma Standard Solution, Specpure®, B 1000 µg/ml, Thermo Fisher Scientific
Standards Cationic Standards Boron Standards
Molecular Formula: H3BO3 in H2O
Physical Form: Liquid
MDL No.: MFCD00134034
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR