Moticam® Pro 205C Digital Microscope Camera, Motic Instruments
1100600120343
89130-164EA 3588.94 USD
89130-164
Moticam® Pro 205C Digital Microscope Camera, Motic Instruments
Motic Images Advanced 3.2 ***Use the camera straight out of the box and start quantifying images or use the programs Z-stacking Multi Focus or Assembly features***Software Development Kit ***Get immediate access to the camera's SDK and make it part of your own software***TWAIN ***Use the camera with third party software using the popular TWAIN interface***DS Compatible *** Make the camera a Direct Show compatible device and use it with an even larger variety of existing third party software***USB2.0 Plug & Play *** Don't search for adaptors, use your laptops integrated USB2.0 for ultimate plug & play convenience***On board Frame Buffer *** Utlize the cameras 4-frame buffer for faster and more consistent frame rates and customize the usage with the SDK package***On board Image Processing ***Get the camera to do some pre-work; use the SDK to adjust and calculate the image before it gets to your computer, do application-specific white balance or color correction straight from the camera***External Trigger Port (TTL) *** Integrate the camera into an event-specific trigger algorithm; the Moticam Pro then becomes even more versatile***Schott BG-40 Filter *** Using this filter allows the camera to pickup only the best nuances of color from the sample
The Moticam Pro 205C Digital Camera uses a C/CS mount to connect to your microscope. The chip size is 1/2” CCD Monochrome with a resolution of 1360 x 1024 Pixels and a pixel sixe of 4.65 x 4.65. This scientific grade camera includes an onboard 4-frame image buffer and a TTL port. Our Pro Software Package CD is included and software updates are free of charge on the Motic website.
Certificaciones: UL Listed.
Información para pedidos: Camera is supplied with all accessories listed above. Contact your VWR representative to learn more about this and other Motic solutions. All Motic products come with a 1 year electrical and 5 year mechanical warranty.