Moticam® Pro 282A Digital Microscope Camera, Motic Instruments
Supplier: Motic
Moticam®
1100600100501
89130-156EA 3070.08 USD
89130-156
Moticam® Pro 282A Digital Microscope Camera, Motic Instruments
Cameras
Capture high-quality images with this scientific-grade CCD color camera.
  • 5.0 megapixel resolution
  • 2580×1944 active pixels
  • Includes software
Internal 4-frame buffer provides faster and more consistent frame rates. On-board image processing allows you to apply white balance or color corrections before the image is sent to your computer. Capture live, still, time-lapse, and video images with the included Motic Images Advanced 3.2 software. Software features include measurement, editing, amalgamation, manual/single point segmentation, single image assembly from multiple images, and multi-language display. USB 2.0 output provides easy communication with your PC or laptop and quick plug-and-play setup.
Certificaciones: UL Listed.
Información para pedidos: Camera is supplied with all accessories listed above. Contact your VWR representative to learn more about this and other Motic solutions. All Motic products come with a 1 year electrical and 5 year mechanical warranty.