Ground & Waste Water Pollution Standard Solution 3, Specpure®

Supplier: Thermo Scientific
Specpure®
36750-AP 036750-AE 36750-AE 36750-AP
AA36750-APEA 303.03 USD
AA36750-AP CAAA36750-AP AA36750-AE CAAA36750-AE
Ground & Waste Water Pollution Standard Solution 3, Specpure®
Standards Environmental Analysis Standards
For US EPA Clean Water Act.

  • 6 Analytes

UN No.: 3264
MDL No.: MFCD02091710
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR