Ceramic Tubes, AL23, Both Ends Open, Thermo Scientific
Supplier: Thermo Scientific
31573-DM 031568-EM 31573-EM 31575-EO 31579-EO 031579-DR 31581-DM 31577-EH 31575-EM 31577-EO 031574-EH 31573-EM 31572-EM 31564-EO 31581-EM 31573-DW 31568-DM 31573-DM 31568-DW 31575-DR 31579-EM 31564-DR 31574-QAT 31573-EH 31564-QAT 31568-QAT 31564-DW 31573-QAT 31575-DM 31577-DR 31574-EM 31564-EM 31581-EO 31568-EO 31581-DW 31577-EH 31564-QAT 31573-EO 031568-EH 31564-DM 31581-DM 31575-DW 31568-EO 31574-DM 31577-EO 031573-DR 31572-EO 31575-EH 31574-EM 31579-EM 31575-EH 31572-DM 31575-DR 31581-EH 31564-EH 31568-DM 31581-DR 31581-DR 31579-DM 31581-EM 31577-DM 31564-DR 031573-EH 031575-DW 31572-DR 31568-DR 31574-DW 31577-DW 31568-QAT 31579-EO 31579-DR 31579-EH 31573-QAT 031581-EH 31572-DM 31568-EH 31572-DW 31568-DW 31573-EO 31579-DW 31581-QAT 31572-EO 31577-EM 31564-EM 31572-EM 031575-EO 31574-DW 31572-EH 31577-QAT 031574-DR 31577-DM 31568-DR 31572-DR 31574-EO 31564-EH 31564-DM 031577-DR 031575-DM 31577-QAT 31564-EO 031564.DW 31572-QAT 31579-QAT 031572-EH 031572-DW 031573-DW 031575-QAT 31577-DW 31568-EM 31579-DW 31574-EH 31572-QAT 31579-QAT 31581-DW 31575-QAT 31573-DR 031579-DM 31581-EO 31574-DR 31575-EM 31574-DM 31577-EM 31574-QAT 31579-EH 31581-QAT 31574-EO
AA31568-EHEA 118.99 USD
AA31568-EH AA31574-EO AA31577-DR CAAA31568-QAT AA31574-EM AA31568-EO AA31568-EM AA31577-DW CAAA31579-DM AA31568-DW CAAA31573-EO CAAA31573-EM AA31568-DR AA31577-QAT AA31577-EM AA31577-EH CAAA31579-EH AA31579-QAT CAAA31574-QAT AA31577-EO AA31572-DM CAAA31579-EO AA31568-DM AA31572-DR AA31572-DW CAAA31579-EM CAAA31579-DW CAAA31579-DR AA31572-EH CAAA31568-DM AA31581-DW AA31572-EM AA31575-DM CAAA31568-DW AA31572-QAT AA31575-DR CAAA31568-DR AA31572-EO AA31575-DW CAAA31577-DM CAAA31574-EH CAAA31574-EM CAAA31568-EH CAAA31581-QAT CAAA31574-DM CAAA31564-QAT AA31581-DM CAAA31574-DR AA31575-EH CAAA31572-QAT CAAA31574-DW CAAA31568-EO AA31581-DR CAAA31568-EM AA31575-EO AA31574-QAT CAAA31577-EH AA31575-EM CAAA31579-QAT CAAA31577-EM CAAA31577-EO AA31581-EH CAAA31574-EO CAAA31577-DW AA31564-EM AA31581-EM CAAA31577-DR AA31581-EO AA31564-EO AA31573-DM AA31573-DR CAAA31577-QAT CAAA31575-DM CAAA31572-EH AA31564-EH AA31573-DW CAAA31572-DM AA31564-DR AA31564-DM CAAA31572-DR AA31568-QAT CAAA31572-DW AA31564-DW AA31573-EH AA31573-EM CAAA31581-EH AA31573-EO CAAA31575-EH CAAA31575-EM CAAA31573-QAT CAAA31581-DW CAAA31572-EO CAAA31572-EM CAAA31575-QAT CAAA31581-DR CAAA31575-DR AA31579-DM CAAA31575-DW CAAA31564-DM CAAA31581-DM CAAA31564-DR AA31579-DW AA31579-DR CAAA31564-DW AA31573-QAT AA31579-EH CAAA31575-EO AA31581-QAT AA31579-EM AA31564-QAT CAAA31564-EM CAAA31581-EO AA31579-EO CAAA31581-EM CAAA31564-EH AA31574-DM CAAA31564-EO AA31574-DR CAAA31573-EH AA31574-DW CAAA31573-DM AA31575-QAT CAAA31573-DR CAAA31573-DW AA31577-DM AA31574-EH
Ceramic Tubes, AL23, Both Ends Open, Thermo Scientific
Tubing Transfer Tubing
Pure Al₂O₃ Aluminium oxide AL23.
  • Corrosion resistant
  • Permeable to microwaves
Dense, excellent thermal and electrical resistance properties.
Outer diameter ranges from 12 to 60 mm.