Anti-MT Mouse Monoclonal Antibody
89359-288
: Genetex
- Antibody Type:Primary
- Antigen Name:Metallothionein
- Antigen Symbol:MT
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Fish
- Western Blot:Yes
- Storage Buffer:Phosphate buffered saline, pH 7.2, PMSF and 50% glycerol
- Storage Temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Immunogen:Full length protein (cross linked rabbit liver MT1 and MT2).
- Tested Applications:WB
- Purification:Protein G purified
- Cat. No.:89359-288
- Supplier no.:GTX12228
Metallothioneins (MTs) are a family of low molecular weight, heavy metal-binding proteins characterized by a high cysteine content and lack of aromatic amino acids. MTs bind 7 to 12 heavy metal atoms per molecule of protein. They are ubiquitous in the animal and plant kingdoms and are also found in prokaryotes. In mammals, the cysteine residues are absolutely conserved and serve to coordinate heavy metal atoms such as zinc, cadmium, and copper via mercaptide linkages.
Type: Primary
Antigen: MT
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Fish