Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:Mineralocorticoid R/NR3C2
- Antigen Symbol:Mineralocorticoid Receptor
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100μl
- Gene ID:4306
- Antigen Synonyms:NR3C2VIT|mineralocorticoid receptor delta|nuclear receptor subfamily 3|MGC133092|mineralocorticoid receptor 1|group C|MRMCRaldosterone receptor|member 2|MLRmineralocorticoid receptor|Nuclear receptor subfamily 3 group C member 2|FLJ41052
- Storage Buffer:PBS and 2% Sucrose
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptides corresponding to NR3C2(nuclear receptor subfamily 3, group C, member 2) The peptide sequence was selected from the middle region of NR3C2. Peptide sequence RRKNCPACRLQKCLQAGMNLGARKSKKLGKLKGIHEEQPQQQQPPPPPPP.
- Purification:Protein A purified
- Cat. No.:102199-890
- Supplier no.:NBP1-70640
Rabbit Polyclonal Mineralocorticoid Receptor Antibody. Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse, Rat, Chicken.
Type: Primary
Antigen: Mineralocorticoid Receptor
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human