The TLR4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TLR4. This antibody reacts with human, mouse. The TLR4 Antibody has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:50
Type: Primary
Antigen: TLR4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Catalog No:
- 102110-138
- 102110-140
- Antigen Symbol:
- TLR4
- TLR4
- Antigen Name:
- Toll-like Receptor 4
- Toll-like Receptor 4
- Antigen Synonyms:
- toll-like receptor 4|CD284 antigen|CD284|ARMD10|TOLL|hTollhomolog of Drosophila toll
- toll-like receptor 4|CD284 antigen|CD284|ARMD10|TOLL|hTollhomolog of Drosophila toll
- Antibody Type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 7099
- 7099
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- A synthetic peptide made to an internal portion of the mouse TLR4 protein (between residues 400-450) [NP_612564]
- A synthetic peptide made to an internal portion of the mouse TLR4 protein (between residues 400-450) [NP_612564]
- Purification:
- Immunogen affinity purified
- Immunogen affinity purified
- Concentration:
- 1.12 mg/ml
- 1.12 mg/ml
- Storage Buffer:
- PBS and 30% Glycerol
- PBS and 30% Glycerol
- Storage Temperature:
- Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- SizeSupplier no.
Specifications
Antigen SymbolTLR4Antigen NameToll-like Receptor 4Environmentally PreferableAntigen Synonymstoll-like receptor 4|CD284 antigen|CD284|ARMD10|TOLL|hTollhomolog of Drosophila tollAntibody TypePrimaryClonalityPolyclonalConjugationUnconjugatedReactivityHuman, MouseHostRabbitGene ID7099IsotypeIgGWestern BlotYesImmunoChemistryYesImmunoFluorescenceYesFlow CytometryYesImmunogenA synthetic peptide made to an internal portion of the mouse TLR4 protein (between residues 400-450) [NP_612564]PurificationImmunogen affinity purifiedConcentration1.12 mg/mlStorage BufferPBS and 30% GlycerolStorage TemperatureStore at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.Specifications
Antigen SymbolTLR4Antigen NameToll-like Receptor 4Environmentally PreferableAntigen Synonymstoll-like receptor 4|CD284 antigen|CD284|ARMD10|TOLL|hTollhomolog of Drosophila tollAntibody TypePrimaryClonalityPolyclonalConjugationUnconjugatedReactivityHuman, MouseHostRabbitGene ID7099IsotypeIgGWestern BlotYesImmunoChemistryYesImmunoFluorescenceYesFlow CytometryYesImmunogenA synthetic peptide made to an internal portion of the mouse TLR4 protein (between residues 400-450) [NP_612564]PurificationImmunogen affinity purifiedConcentration1.12 mg/mlStorage BufferPBS and 30% GlycerolStorage TemperatureStore at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.