Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:E. Coli
- Antigen Symbol:E. Coli
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Bacteria
- Size:0.5 ml
- Antigen Synonyms:Escherichia coli
- Storage Buffer:Phosphate buffered saline
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:4.0 mg/ml
- Immunogen:A mixture of E.coli serotypes
- Purification:IgG purified
- Cat. No.:102113-778
- Supplier no.:NB100-64447
The E. Coli Antibody from Novus Biologicals is a goat polyclonal antibody to E. Coli. This antibody reacts with bacteria. The E. Coli Antibody has been validated for the following applications: Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: E. Coli
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Bacteria