Ubbelohde DIN Series Capillary Viscometers, Fungilab
Supplier: Fungilab
CV003-107 CV003-114 CV003-109 CV003-101 CV003-111 CV003-108 CV003-100 CV003-113 CV003-104 CV003-105 CV003-103 CV003-110
89236-870EA 347.23 USD
89236-870 89236-860 89236-872 89236-862 89236-874 89236-852 89236-854 89236-866 89236-878 89236-868 89236-858 89236-880
Ubbelohde DIN Series Capillary Viscometers, Fungilab
Viscometers Capillary Viscometers
Ubbelohde Viscometers are used to determine kinematic viscosities of transparent Newtonian liquids such as jet and hydraulic lubricants.
Certificaciones: UNE 400313, DIN 51562 Part 1 and ISO 3105.
Información para pedidos: Includes constant value calibration certificate at 40ºC (104ºF) and 100ºC (212ºF).