Order Entry
Ammonium heptamolybdate USP Test Solution (TS)
Ammonium heptamolybdate USP Test Solution (TS)
undefined
Ammonium heptamolybdate USP Test Solution (TS)
  • Appearance:
    Clear to hazy, colorless to light green with slight nitric acid odor
  • Suitability for Use:
    Yellow precipitate with Phosphate
  •  
    Pack type
    Size
    Supplier no.
  • RC674-4
    R0674000-120B
    Poly bottle, amber
    120 ml
    R0674000-120B
    Each (120ml)
    Each (120ml) - $89.00
    $89.00
    Each (120ml)
       $89.00
    Small Business Enterprise
    Small Business Enterprise
  • RC674-16
    R0674000-500B
    Poly bottle, amber
    500 ml
    R0674000-500B
    Each (16oz)
    Each (16oz) - $186.00
    $186.00
    Each (16oz)
       $186.00
    Small Business Enterprise
    Small Business Enterprise