EnviroMax® Environmental Sampling Swabs, Round Foam Tip, Puritan Medical Products
Supplier: Puritan Medical Products
88010-PFRND 25-88010PFBRND 25-88010PFRND 88010-PFBRND
89221-736CS 829.17 USD
89221-736 89221-738 89221-734 89221-740
EnviroMax® Environmental Sampling Swabs, Round Foam Tip, Puritan Medical Products
Swabs Swabs for Sampling
These kits are an excellent choice for sample collection on large surfaces or equipment parts
The benefits of foam-tipped swabs are the large tip, which increases sample and elution counts, and the rigid paddle-like design which allows for the firm and even application of pressure necessary for adequate sample collection. To use, simply remove Puritan's EnviroMax® swab from the vial, collect the sample, and return the swab to the vial.
Shelf life is 12 months. Made in USA.
The oversized swab consists of a rigid paddle inside a foam tip. The swab tip is made out of a medical-grade polyurethane foam and is securely attached to a polypropylene handle. Tips come in two configurations; pointed for hard-to-reach areas such as drains, or round for use on floors, walls, and countertops.
Información para pedidos: Swabs are packaged 25 individually wrapped tubes per case. Bulk Packaging includes 10 boxes of 25 individually wrapped packages per box.