HyClone SFMTransfx-293 is a serum-free, animal derived component-free medium designed to support the growth of HEK 293 cultures and promote transfection using lipofection or similar methods. Manufactured according to cGMP guidelines.
- Supports high transfection efficiency
- Metabolically designed for high cell yield and recombinant protein/vector production
- Allows for direct or sequential adaptation
HyClone SFMTransfx-293 Media is a regulatory-friendly medium developed through the Metabolic Pathway Design to support high transfection efficiency, productivity and cell density in suspension cultures.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Storage Temperature:2...8 °C
- Media FormatSizeSupplier no.
- SFMTranfx-293 medium without L-glutamine, phenol red, or HEPES, with poloxamer 188 and sodium bicarbonateSFMTranfx-293 medium without L-glutamine, phenol red, or HEPES, with poloxamer 188 and sodium bicarbonateSH30860.02Liquid1 LSH30860.02Each (1,000ml)$127.51
- SFM4Transfx-293 medium without L-glutamine, phenol red, HEPES, sodium bicarbonateSFM4Transfx-293 medium without L-glutamine, phenol red, HEPES, sodium bicarbonateSH30861.01Powder5 LEach (5l)$307.68