HyClone™ Non-Essential Amino Acids, HyClone products (Cytiva)
16777-186
: Cytiva
:
Specifications
- Description:NEAA solution, 100X
- Size:100 ml
- Cat. No.:16777-186
- Storage temperature:+2…+8 °C
- Volume:Ea. (100ml)
- Supplier no.:SH30238.01
Specifications
HyClone Amino Acids are ideal for supplementation of serum- and protein-free media
They Support the growth of cells that have high energy demands and synthesize large amounts of proteins and nucleic acids.