Puerto Rico
Order Entry
(9H-Xanthen-9-yl)acetic acid
  101797-744
 :  
undefined
(9H-Xanthen-9-yl)acetic acid
Catalog # 101797-744
Supplier:  Matrix Scientific
Supplier Number:  010035-500MG
CAS Number:  
Price
$267.69
Log in for availability
Unit of Measure Each (500mg)

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Size:
    500 mg
  • Cat. No.:
    101797-744
  • Supplier no.:
    010035-500MG

 

 

 :
 :240.26 g/mol
 :155…156 °C
 :1217-58-9
 :C₁₅H₁₂O₃
 :MFCD00197336