HyClone® Insect Cell Screened Fetal Bovine Serum, U.S. Origin
SH30070.03I
82007-420EA 1201.89 USD
82007-420
HyClone® Insect Cell Screened Fetal Bovine Serum, U.S. Origin
HyClone Insect Cell Screened Fetal Bovine Serum, U.S
  • Highest quality serum
  • Support robust growth of insect cells
  • U.S. sourced
HyClone Insect Cell Screened FBS starts with Defined Serum, the highest quality available. Test lots are compared to a control lot that has demonstrated the ability to support robust growth of insect cells. Storage Temperature is -10°C.
Origin is ready-to-use and prescreened to identify lots that offer optimal performance with BEVS.