Specially designed for use in the laboratory, these rugged and durable benchtop meters are versatile enough to fit your unique testing needs.
- Simple to operate and maintain
- Consistently accurate and reliable measurements
- Configurable to each user's unique needs
- VWR® three-year limited warranty
- Available with ISO/IEC 17025 NIST traceable calibration certificate
These meters are simple to operate and offer an intuitive user interface allowing for easy set up and use from day one.
Calibrated pH meters and kits include a Troemner ISO/IEC Accredited Calibration Certificate traceable to NIST. The ISO/IEC 17025 Accredited Calibration Certificate includes electrical simulation of multiple data points within the functional ranges with the associated uncertainties. Troemner is accredited by NVLAP under Laboratory Code 105013-0 to perform ISO/IEC 17025 calibrations.
Step-by-step on screen prompts enable both new and seasoned users to operate the meter easily and effectively, resulting in consistently accurate measurements.
Certificaciones: VWR® sympHony™ benchtop meters meet IP54 specifications for protection from water and are CE and cETLus marked.
Información para pedidos: Stand is not included with the meter kits. Each benchtop meter can be configured to fit your needs with the variety of optional electrodes, stands with stirrers, and accessories available.