Copper ICP Standard, 1 ml = 1 mg Cu (1000 ppm Cu)Cu in 3% HNO₃, Ricca Chemical Company
Supplier: RICCA Chemical Company
RPCU1KN0-100N RPCU1KN0-500N
RCPCU1KN-500EA 292.93 USD
RCPCU1KN-500 RCPCU1KN-100
Copper ICP Standard, 1 ml = 1 mg Cu (1000 ppm Cu)Cu in 3% HNO₃, Ricca Chemical Company
Standards Cationic Standards Copper Standards