Bar Code Readers, WHEATON
Supplier: DWK Life Sciences
W986010 W986000
89232-016EA 14185.24 USD
89232-016 89194-788
Bar Code Readers, WHEATON
The WHEATON SingleScan and PluraScan offer versatility, with easy set up
The WHEATON SingleScan offers plug and play versatility, with easy set up and no software installation required. The USB interface allows easy connection to any computer. It is capable of reading both 1D linear bar codes and 2D Data Matrix bar codes on any vial or ampule. The decoded bar code ID is placed into any software application where the cursor is placed.
The WHEATON PluraScan Bar Code Reader makes sample traceability easy. The PluraScan Bar Code Reader is a sophisticated 2D Data Matrix Bar Code Reader capable of reading bar codes from a wide range of manufacturers and label printers. The PluraScan Bar Code Reader is out of the box compatible with the WHEATON Keep-It® 81, & 100 Boxes for use with the WHEATON CryoELITE cryogenic vials. The PluraScan is versatile and can be adapted for use with multiple different rack and box formations.
Operating System for 89194-788 are Factory configured for Windows® operating systems compatibility. Also compatible with non-windows operating systems. For 89232-016 are Windows® 7, 2000, XP.
Capable of reading both 1D linear bar codes and 2D Data Matrix bar codes on any vial or ampule.