Corning ® Glass Fiber Prefilters
Supplier: Corning
Corning®
431412 431410 431413 431411
97039-894CS 85.57 USD
97039-894 97039-896 97039-898 97039-900
Corning ® Glass Fiber Prefilters
Filters Glass Fiber Filters
Glass fiber filters are used a depth filter for prefiltration of solutions.
They have very high particle loading capacity and are ideal for prefiltering dirty solutions and difficult to filter biological fluids such as sera.