Corning® 384-Well Low Volume Microplates
Supplier: Corning
3821 3822 3826 3825 3820 3824
89089-858CS 783.32 USD
89089-858 89089-856 89089-868 89089-870 89089-860 89185-546
Corning® 384-Well Low Volume Microplates
Microplates Deep Well Microplates
Corning 384 well solid black and white polystyrene microplates are designed to reduce well-to-well crosstalk during fluorescent and luminescent assays.