SteamScrubber® 33 Glassware Washer, Labconco
4578030 4578020 4578031 4578021
89232-216EA 12342.47 USD
89232-216 89232-214 89232-212 89232-210
SteamScrubber® 33 Glassware Washer, Labconco
SteamScrubber® 33 Glassware Washers are designed to wash and dry general-purpose labware
They have stainless steel top and bottom racks that accommodate basket inserts for a wide variety of glassware shapes and sizes, primarily beakers and other wide-mouth glassware. Specialized inserts for BOD bottles, petri dishes, culture tubes, and small utensils enhance the unit's versatility. In addition, the SteamScrubber® accommodates upper and lower Spindle Racks. (Additional racks and basket inserts are sold separately). Minimum height allows washer to fit under ADA-height cabinetry. Undercounter model.