PYREX® Capillary Tubing
236520
470174-212
PYREX® Capillary Tubing
Tubing Transfer Tubing
Made from high quality borosilicate with inside diameter tolerances of ±0.15 mm and wall tolerances of ±0.15 mm. The larger sizes can be used as barometer tubes. Supplied in 150 cm lengths and sold by the pound.