SP Bel-Art Earth-Friendly Wide Handle Sampling Spoons, Bel-Art Products, a part of SP
Supplier: Bel-Art
SCIENCEWARE®
F36740-1024
89218-778CS 204.65 USD
89218-778
SP Bel-Art Earth-Friendly Wide Handle Sampling Spoons, Bel-Art Products, a part of SP
Spoons
Lessen environmental impact and increase contributions to sustainability goals with these PLA (polylactic acid) resin sampling spoons.
  • PLA resin construction
  • Earth-friendly with no petroleum-based materials
  • For retrieving small, specific volume samples
  • Nonsterile
The production of 100% plant-based PLA contributes less greenhouse gasses and uses less nonrenewable energy from source to resin than its petroleum-based counterparts, and the plants used are an annually renewable resource.
Disposal options include industrial composting for biodegradability, incineration, recycling, and landfill. Safe for food, drug, and cosmetic contact, these FDA-grade spoons are comparable in strength and durability to polystyrene, polypropylene, and other plastic resin sampling spoons.
Perfect for retrieving small, specific volume samples of powders or liquids. The wide handles enable a firm grip and provide just enough stiffness to easily sample packed powders and other firm materials. Their unique flat-bottom shape allows convenient dual use as weighing boats to save time and assure accuracy. Use a separate spoon for each material sampled to avoid cross contamination. Nonsterile.