You Searched For: Water Bath Protection Reagents
Since the moist and warm environment is perfect for microbe growth, water baths are often susceptible to contamination which is the biggest reason testing results become skewed. Using water bath protection reagents though will reestablish the hygienic conditions for stable incubation, heating, and thawing. When the correct ratio is used, the water bath protection reagents will successfully prevent algae, fungi, and bacteria from forming keeping water clear for weeks.
2 results were found
Sort Results
List View
Easy View