CBS Scientific offfers a choice of 3 power supplies to cover a wide range of electrophoresis applications.
- All 3 types of power supplies have the same compact, light and stackable design
- Universal input power supply accepts all domestic and international voltages
- Parameters are set by pressing UP/DOWN arrow buttons in combination with a MODE button
- Power output is controlled by setting a maximum value for voltage or current
Safety features on all three models include no load detection, sudden load change protection, over voltage protection and automatic power failure recovery.
The power supply automatically “crosses over” or switches the limiting parameter according to the programmed limits as the resistance of the electrical load changes during a run.
Certificaciones: CE marked.