You Searched For: Color Reference and Spectroscopy Standards
Color reference and spectroscopy standards are used to routinely monitor optical spectrometer mechanisms. The calibration solutions provide the base readings that users can compare with the electromagnetic radiation wavelengths and intensities scientific instruments obtain. Placing the reference solution inside, users can zero out settings to reestablish proper mechanics and eliminate systematic error. For quality control, the color reference and spectroscopy standards guarantee that the machines function properly for accurate spectral lines.
74 results were found
Sort Results
List View
Easy View