Would you like to log in again?
VWR® Floor Model Shaking Incubator
Supplier: VWR International
1585-2 1585R-2 1585 1585R
97051-682EA 10169.58 USD InStock
97051-682 97051-686 97051-680 97051-684
VWR® Floor Model Shaking Incubator
Shakers and Mixers Shaking Incubators
Floor shaking incubators feature a unique, adjustable counterbalance weighting system to accommodate off-center loads and varying capacities and stroke lengths.
  • Overtemperature protection
  • Features include controls for temperature, timer, and shaker
  • Small footprint
An adjustable orbit allows users to change the rotational stroke from vigorous shaking to gentle shaking to accommodate different types of cells, and to provide maximum oxygen transfer.
Model 1585 has a small footprint and its chamber provides shelf space for static incubation during shaking. The 1585R refrigerates via a 1/6-horsepower motorized compressor (produced without CFCs/HCFCs).
Standard features include microprocessor control, load flexibility, shaking platform (included), and independent controls for temperature, timer, and shaker.
All major functions (temperature, RPM and time) have alarms that alert the user to deviations from set parameters. Overtemperature protection is provided by a safety thermostat. Digital keypad operation offers the ability to calibrate the temperature controller to a reference thermometer. To better support the load over years of constant use, four load bearing positions for optimal weight distribution are provided.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
SERVICIOS
Servicios: VWRCATALYST™ offers customer on-site Calibration, Preventative Maintenance and validation services including IQ, OQ, and PQ protocol development and execution services (US only).Please Call 888-793-2300 or visit www.vwr.com/equip_services for more details.
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty