You Searched For: Forensic Enclosures
Forensic enclosures provide a contamination-free environment when maintaining the integrity of evidence is paramount. Incorporating filters and a ventilation system for personnel safety, the chambers use fuming methods to protect crime scene materials. The hinged glass doors allow superior visibility during fingerprint fixing or monitoring fuming cycles. With additional features to guarantee maximum preservation during storage, the forensic enclosures have tamper-free locking capabilities and ultraviolet light disinfection.
4 results were found
Sort Results
List View
Easy View