CAMCO® Guaiac-Tabs For Qualitative Determination of Fecal Occult Blood, Cambridge, Cambridge
Supplier: Cambridge
Camco®
1003
CACM1003
CAMCO® Guaiac-Tabs For Qualitative Determination of Fecal Occult Blood, Cambridge, Cambridge
Clinical Diagnostic Supplies Clinical Reagents and Kits
Guaiac-Tabs is a simple reliable test that provides a sensitive and convenient test for qualitative determination of occult blood in feces
Guaiac Tabs provide a Quick, simple procedure with results in 2 minutes. 100 filter paper squares provided.
The specimen is placed on filter paper; then a tablet is placed in the center of the specimen. Add two drops of glacial acetic acid and two drops of 3% hydrogen peroxide to the tablet. Filter paper turns blue if the test is positive. If blue color does not appear within two minutes, the test is negative.
Información para pedidos: 100 Tablet bottle and 100 pieces of filter paper provided. You will need glacial acetic acid and 3% hydrogen peroxide to complete the procedure.