CONTACT SLIDE CHROM 1 120SLIDES PK120
Supplier: LIOFILCHEM, INC.
Contact Slide Chrom 1, Chromatic Coli Coliform/Chromatic Salmonella, Flex Dip-slide with chromogenic selective media for detection of Escherichia coli, other coliform bacteria and Salmonella spp, rtu device with 2 different media coated onto plastic support used for microbial monitoring, 120slides
Learn more About VWR
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com