Hoshizaki C-100BAF-AD Ice Maker with Bin, Nordon Inc.
C-100BAE-AD
89166-540EA 6975.15 USD
89166-540
Hoshizaki C-100BAF-AD Ice Maker with Bin, Nordon Inc.
The C-100BAF-AD self contained cubelet design is small enough to fit conveniently into most cabinetry without additional ventilation
The swing style reversible door and the UL outdoor approval expand the possibilities where the unit can be installed. It produces up to 92 lbs of ice per 24 hour period with up to 29 lbs of built in storage. It has fewer moving parts for a longer life. The power swith is easily accesible without having to remove the panel. You can easily disassembly the water circuit for easy cleaing. This unit complies with ADA standards
Certificaciones: Ice maker is ETL, NSF certified and UL listed. Filters are AHRI and NSF certified.
Información para pedidos: Ice maker is supplied with manufacturer's three-year warranty on parts and labor and five-year warranty on compressor and air cooled condenser coil. 5 Year warranty on heads offered by Everpure.