The KM Cuber Series features an attractive stainless steel finish with the simplest icemaking system in the industry
It produces up to 88 lbs of ice per 24 hour period with up to 53lbs of built in storage. The exclusive dual-sided stainless steel evaporator has an oval-shaped copper refrigerant tubing for the most efficient heat exchange. It's fewer moving parts and cycles which means dependability, longer machine life and energy savings. The KM models also feature Cyclesaver which allows the same amount of ice in half of the cycles as other machines. It has an EverCheck control board which sounds an audible warning when service is needed. This unit also contains HoshiGuard Antimicrobal Agent.
Certificaciones: Ice maker is AHRI, ETL and NSF certified. Filters are AHRI and NSF certified.
Información para pedidos: Ice maker is supplied with manufacturer's three-year warranty on parts and labor and five-year warranty on compressor and air cooled condenser coil. Filters are supplied with five-year manufacturer's warranty on heads offered by Everpure.