VWR Life Science Bluestep™ Protein Molecular Weight Marker, Low Range, Prestained
BLUESTEP™
K972-0.5ML
97063-714EA 232.64 USD
97063-714
VWR Life Science Bluestep™ Protein Molecular Weight Marker, Low Range, Prestained
  • Electrophoresis Grade
  • 6 bands ranging from 19.0 to 120.0 kDa
  • Pre-stained bands are easily transferred to blotting membranes for use in Western blotting
  • Monitor real-time running conditions during electrophoresis