- Sensitive, fast and consistent protein assay kit
- Determine protein concentrations from 1 - 100 μg
The Bradford Protein Assay is the preferred colorimetric assay for quantifying total protein concentration. Based upon complex formation between basic and aromatic amino acid residues with Coomassie® Brilliant Blue G-250 dye1, the Bradford method is easier, faster and more sensitive than the Lowry method. The standard protocol is useful for measuring between 10 - 100 μg of protein and the microassay is useful for measuring between 1 - 10 μg of protein. Kit includes: Bradford Reagent, 1 L and Albumin, Bovine - 0.5 mg/ml Standard, 1.5 ml, Sodium Chloride 0.15M, 200 mL