You Searched For: Glassware Washers
Professional washing and thermal disinfection machines are manufactured using high-quality materials and components. These front-loading stainless steel devices can fit under work surfaces and have a large washing compartment with multiple levels. The appliances clean common fragile pieces in industry, environmental, and research laboratories to analytically pure standards. Glassware washing machines are the perfect choice to wash and disinfect all instruments including laboratory glassware, test tubes, narrow neck glassware, used in small laboratories, schools, or specialized clinics. FlaskScrubber Glassware Washers are designed to wash and dry narrow-neck and general purpose labware. Specialized inserts for pipettes, BOD bottles, petri dishes, culture tubes and small utensils make the FlaskScrubber versatile.
22 results were found