You Searched For: Indicator Paper

Ready for immediate dip and read processing, indicator papers determine the contents of a sample through simple color comparisons with supplied charts. Saturated with a mixture of several reagents, the fast-acting quantitative detectors provide accurate colorimetric answers. The sharp color appearance facilitates routine pass-or-fail assessments. While universal litmus indicator papers provide acidity or alkalinity readings, graduated tests give specific element levels. Use these compact tests in the laboratory or on-site.

Ready for immediate dip and read processing, indicator papers determine the contents of a sample through simple color comparisons with supplied charts. Saturated with a mixture of several reagents, the fast-acting quantitative detectors provide accurate colorimetric answers. The sharp color appearance facilitates routine pass-or-fail assessments. While universal litmus indicator papers provide acidity or alkalinity readings, graduated tests give specific element levels. Use these compact tests in the laboratory or on-site.


23  results were found

SearchResultCount:"23"

Sort Results

List View Easy View

Rate These Search Results

Catalog Number: (EM1.09512.0003)
Supplier: MilliporeSigma
Description: Potassium iodine paper is used for the determination of oxidizing agents.

Catalog Number: (89500-724)
Supplier: REACTIVOS Y EQUIP SA DE CV MX
Description: Vial of 100 test strips.


Catalog Number: (89231-118)
Supplier: REACTIVOS Y EQUIP SA DE CV MX
Description: Hydrogen Sulfide Test Papers, for Test Kit HSC.

SDS Product available on GSA Advantage®


Catalog Number: (90002-220)
Supplier: BD
Description: Taxo™ ONPG Discs are used for the detection of lactose fermenters, especially those that do not promptly ferment lactose in some routine identificatio...

Catalog Number: (90002-462)
Supplier: BD
Description: Paper Discs for the Detection of Beta-Lactamase Enzymes

Supplier: Mar Cor Purification
Description: Minncare 1% Test Strips verify adequate concentration of a 1% Minncare solution during sanitization of your high purity water system
Catalog Number: (66170-324)
Supplier: LaMotte
Description: Test paper for the determination to test iodine in water.

SDS


Supplier: Precision Laboratories
Description: Thiourea Test Paper, Vial of 100

Catalog Number: (76579-374)
Supplier: Mortech Manufacturing
Description: Test Strips for use with Formaway Formalin Neutralizing products


Catalog Number: (89370-914)
Supplier: Micro Essential
Description: A quick, reliable, easy-to-use, way to test quaternary ammonium based sanitizers in food service establishments, as required by federal, state, and lo...


Catalog Number: (470123-112)
Supplier: Precision Laboratories
Description: Cobalt Chloride Test Paper


Supplier: Cytiva
Description: Cytiva offers a range of pH indicator and test papers to meet specific needs
Catalog Number: (10217-568)
Supplier: Macherey-Nagel
Description: Quantofix Lubricheck test strip for the semi-quantitative determination of the concentration of cooling lubricants


Catalog Number: (470023-872)
Supplier: Kemtec
Description: Standard hemoglobin reference scale.


Catalog Number: (66170-326)
Supplier: LaMotte
Description: Chlorine test papers are chemically treated to indicate sanitizer levels.

SDS Small Business Enterprise


Catalog Number: (470093-682)
Supplier: Precision Laboratories
Description: A simple dip test for presence of lipids in solution.


Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 16 of 23
no targeter for Bottom