Sample Containers, Polypropylene, Greiner Bio-One
Supplier: Greiner bio-one
724402 724461
89085-302CS 267.2 USD
89085-302 89085-304
Sample Containers, Polypropylene, Greiner Bio-One
Sample Containers General Purpose Containers
Multipurpose polypropylene containers provide unbreakable, leak-proof security for sample storage and transport.
  • Polypropylene construction
  • Unbreakable and leakproof
  • Containers are autoclavable
  • Supplied with HDPE screw cap
Containers offer high thermal, mechanical, and chemical resistance, and feature graduation marks and a writing space
For improved traceability, catalog number, lot number, and expiration date are printed on the base of each container. Designed for single use only. Containers are autoclavable. Screw caps are not. Dimensions: 7.2Lx6.2Dia. cm (227/32x27/16"). Color: natural.
They are also equipped with an integrity seal to ensure sterility and intactness. All containers are supplied with an HDPE screw cap.