Nunc® Lab-Tek™ Multi-Purpose Lab Containers, Thermo Scientific
Supplier: Thermo Scientific
Nunc® Lab-Tek™
4713 4714 4705 4713 4714 4704 4715 4704 4715 4705
15707-952CS 152.62 USD
15707-952 CA15707-954 CA15707-964 15707-966 15707-954 CA15707-952 15707-964 CA15707-968 CA15707-966 15707-968
Nunc® Lab-Tek™ Multi-Purpose Lab Containers, Thermo Scientific
Sample Containers General Purpose Containers
Containers used for collection, transport, and storage of lab specimens.
Durable polypropylene containers are easy-open, tight-sealing, resistant to most chemicals, including formaldehyde, also withstand temperature extremes from freezing to boiling.
Polystyrene containers withstand high impact, have better rigidity, are more stackable, and have an easy write-on surface.
High-density polyethylene containers with lid are reliable and durable, nonsterile.