96-Well Clear Bottom Plates, Corning®
Supplier: Corning
3610 3603 3904 3631 3632 3604 3995 3903 9102 3651
29444-099CS 1370.79 USD
29444-099 89091-018 33501-820 89091-012 29444-008 89091-010 29444-010 29445-162 89091-014 29442-072
96-Well Clear Bottom Plates, Corning®
Microplates Cell Culture Plates
These 96-well flat-bottom polystyrene plates have opaque walls to prevent well-to-well crosstalk and optically clear bottoms to permit direct microscopic viewing
Tissue culture treated plates are sterile, have a cell growth surface area of 0.32 cm2 per well. Nonbinding surface (NBS®) treated plates utilize a nonionic hydrophilic surface (polyethylene oxide-like) to minimize molecular interactions. Non-treated and high binding plates are ideal for assays where visualization is preferred, but cell attachment is not required. Rigid styrene lids available separately.
Plates can be used in both top and bottom reading instruments. Bottoms are 60% thinner than those of conventional plates, resulting in lower background fluorescence and enabling readings down to 340 nm. Plates have a total well volume of 360 µL. Black pigment walls have low background fluorescence and minimal light scatter. Dense white pigment walls enhance luminescent signals, have low background luminescence, and low background fluorescence.
Información para pedidos: Small-pack 360 µL plates are packaged 5 plates per pack, 5 packs per case.