These steel cabinets are specifically designed for the storage of corrosive chemicals.
- Light neutral finish
- LDPE liner
- Nylon roller catch
Cabinets are lined with a molded one-piece, linear low-density polyethylene tub with covered corners and a 2.5 cm (1") lip at the bottom front. The cabinet doors are lined with a 0.3 cm (1/8") polyethylene sheet and feature a nylon roller catch. Each cabinet is furnished with a 5.1 cm (2") diameter PVC vent pipe for venting to the fume hood above and requires a 5.1 cm (2") hole in the hood worksurface. Cabinets also feature doors with radiused full-width integral pulls. Finish is a light neutral color.
Información para pedidos: These REDISHIP items can be shipped within 48 hours of receiving your order. Because restrictions may vary, VWR recommends referencing your local building code requirements when selecting a flammable or solvent base cabinet to ensure you are in full compliance with regional regulations.