Wipe Test Smears, Biodex Medical Systems
006-350
21908-872PK 252.78 USD
21908-872
Wipe Test Smears, Biodex Medical Systems
Sample any surface contaminated with radioactivity
  • Sample any surface contaminated with radioactivity
  • Convenient, easy-to-use cloth discs
  • Individual record folders to perform wipe tests as indicated in NRC and Agreement State Regulations
These convenient, easy-to-use cloth discs have individual record folders and can be used to perform wipe tests as indicated in NRC and Agreement State Regulations. Each box contains 500 record folders which can be used to identify each sample smear, prevent cross-contamination of smears, transfer smears to the counting facility, or store smears until discarded. Smears are 4.4cm (13/4") diameter cloth discs.