Would you like to log in again?
AcroPrep™ 24 Filtration System, Pall Laboratory
AcroPrep™
289000159 289000159
28143-020PK 1467.12 USD InStock
28143-020 CA28143-020
AcroPrep™ 24 Filtration System, Pall Laboratory
Manifolds
The AcroPrep™ filtration system allows simultaneous filtration of 24 samples in only seconds.
  • Accelerate sample preparation
  • HPLC certified
  • Individually sealed wells
Designed to accelerate sample preparation, it saves 20 minutes of labor per carousel over the conventional syringe filter technique
The AcroPrep™ 24-well filter plates feature polypropylene housing with a choice of GH Polypro (hydrophilic polypropylene), nylon, PTFE, or PVDF membranes.
The system can be used with conventional 12x32mm vials, and requires less bench space. HPLC certified to ensure that analytical results will not be compromised by extractable filter materials. Each 1.9mL well is individually sealed to eliminate cross-contamination. The AcroPrep™ system operates by vacuum via conventional lab pumps.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty