LAMBDA™ 25/35 Series UV/Vis Spectrophotometers, PerkinElmer
LAMBDA™
L6020061 L6020067 L6020062 L6020060 L6020065 L6020066 L6020064 L6020063
CA97024-424 CA97024-436 CA97024-430 CA97024-422 CA97024-428 CA97024-434 CA97024-426 CA97024-432
LAMBDA™ 25/35 Series UV/Vis Spectrophotometers, PerkinElmer
Ideal for use in regulated industries and when a wide range of sampling options and large number of samples are required, these spectrophotometers deliver trustworthy results with a minimum of operator training.
  • Double-beam operation
  • Excellent stability
  • Consistent performance throughout lifespan
  • Easy to use
  • Simple replacement of prealigned halogen lamps
True double-beam operation provides the best possible stability and allows references to be measured and corrected in real time
Model 25 is a fixed-bandwidth system and is ideal for most routine UV/Vis applications and regulatory tests. It features a 1nm SBW configuration and meets all pharmacopeia requirements. Cell changers allow for high volume sampling and cell holders are Peltier controlled. For very high volume sampling, a sipper and autosampler are available.
Model 35 features a variable bandwidth that offers the best solution for QC/QA on liquids, solids, pastes, and powders. This model has all of the features of Model 25, but with the flexibility of user selectable bandwidths, from 0.5 to 4nm in four steps. An integrating sphere allows for measurements on cloth, powders, and other diffuse or light-scattered samples.
UV WinLab™ software (included) guides the user through a step-by-step process that simplifies analysis, from sample entry to report generation. For FDA-regulated industries, the optional Enhanced Security™ (ES) version of UV WinLab™ integrates seamless 21 CFR Part 11 technical compliance.
Información para pedidos: All models require a PC for operation and are available with or without USA PC systems. Supplied with UV WinLab™ software and a set of two sample compartment windows. For additional accessories, contact your VWR representative for more information.