United States
Order Entry
Oxalate Standard: C2O4(-2) @ 1000 µg/mL in H2O, VHG, LGC Standards
Oxalate Standard: C2O4(-2) @ 1000 µg/mL in H2O, VHG, LGC Standards
New Product
 
undefined
Oxalate Standard: C2O4(-2) @ 1000 µg/mL in H2O, VHG, LGC Standards
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Environmentally Preferable:

 

 

  •  
     
    Size
    Concentration
    Matrix
     
     
  • Oxalate Standard: C2O4(-2) @ 1000 µg/mL in H2O
    Oxalate Standard: C2O4(-2) @ 1000 µg/mL in H2O
    77697-584
    VHG-IOXAL-100
    100 ml
    1000 µg/mL
    H₂O
       
       
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $71.40
    $71.40
    Each(1/Itm/Bx)
       $71.40

     
    New Product
    New Product
  • Oxalate Standard: C2O4(-2) @ 1000 µg/mL in H2O
    Oxalate Standard: C2O4(-2) @ 1000 µg/mL in H2O
    77697-585
    VHG-IOXAL-500
    500 ml
    1000 µg/mL
    H₂O
       
       
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $150.50
    $150.50
    Each(1/Itm/Bx)
       $150.50

     
    New Product
    New Product