Lubricating Oil Standard 11: Ca @ 0.050; P @ 0.150; S @ 0.200; Zn @ 0.200 wt% in Lubricating Oil, VHG, LGC Standards
77697-649
New Product
:
- Concentration:Varies
- Matrix:oil
- Description:Lubricating Oil Standard 11: Ca @ 0.050; P @ 0.150; S @ 0.200; Zn @ 0.200 wt% in Lubricating Oil
- Size:100 ml
- Cat. No.:77697-649