Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
/
Puerto Rico
Products
Services
Digital Solutions
Applications
Support
About
$0.00
Order Entry
Home
Standards
Organic Standards
n-Nonane, Dr. Ehrenstorfer, LGC Standards
n-Nonane, Dr. Ehrenstorfer, LGC Standards
n-Nonane, Dr. Ehrenstorfer, LGC Standards
77678-448
:
LGC Standards
New Product
:
n-Nonane, Dr. Ehrenstorfer, LGC Standards
77678-448
:
LGC Standards
:
DRE-C15623000
New Product
:
$51.00
Each(1/Itm/Bx)
-
+
CAS No.:
111-84-2
Description:
n-Nonane
Size:
1 ml
Formula:
H₃C(CH₂)₇CH₃
Molecular Weight:
128.26 g/mol
Cat. No.:
77678-448