Anti-GJC1 Rabbit Polyclonal Antibody
New Product
ELISA:1:2000-1:10000, WB:1:1000-1:5000
Type:
Antigen: GJC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
- Catalog No:
- 77714-970
- 77714-969
- Antigen Symbol:
- GJC1
- GJC1
- Antigen Synonyms:
- Gap junction gamma 1 protein|GJC1|CX 31.9|Gap junction alpha 7 protein|Connexin-45|Connexin 45|Gap junction gamma-1 protein|Gap junction protein alpha 7|DKFZp686P0738|GJD3|CXG1_HUMAN|Connexin 45 / GJA7|gamma 1|Gap junction protein|GJA7|Gap junction protein alpha 7 45kDa|CX 45|Connexin45|Gap junction alpha-7 protein|Cx45|GJA 7|CX31.9|45kDa
- Gap junction gamma 1 protein|GJC1|CX 31.9|Gap junction alpha 7 protein|Connexin-45|Connexin 45|Gap junction gamma-1 protein|Gap junction protein alpha 7|DKFZp686P0738|GJD3|CXG1_HUMAN|Connexin 45 / GJA7|gamma 1|Gap junction protein|GJA7|Gap junction protein alpha 7 45kDa|CX 45|Connexin45|Gap junction alpha-7 protein|Cx45|GJA 7|CX31.9|45kDa
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P36383
- P36383
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human GJC1
- Synthetic peptide of Human GJC1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
- SizePackaging
- New Product
Specifications
Antigen SymbolGJC1Environmentally PreferableAntigen SynonymsGap junction gamma 1 protein|GJC1|CX 31.9|Gap junction alpha 7 protein|Connexin-45|Connexin 45|Gap junction gamma-1 protein|Gap junction protein alpha 7|DKFZp686P0738|GJD3|CXG1_HUMAN|Connexin 45 / GJA7|gamma 1|Gap junction protein|GJA7|Gap junction protein alpha 7 45kDa|CX 45|Connexin45|Gap junction alpha-7 protein|Cx45|GJA 7|CX31.9|45kDaClonalityPolyclonalConjugationUnconjugatedReactivityHuman, Rat, MouseHostRabbitGene IDP36383IsotypeIgGWestern BlotYesELISAYesImmunogenSynthetic peptide of Human GJC1FormLiquidPurificationAntigen affinity purifiedStorage BufferPBS, 0.05% sodium azide, 40% glycerol, pH 7.4.Storage TemperatureUpon receipt, store at –20 or –80 °C. Avoid repeated freeze.Shipping TemperatureShipped on blue iceNew Product - 77714-969A48607-100UL100 µlVialEach(1/Itm/Bx)$334.95New Product
Specifications
Antigen SymbolGJC1Environmentally PreferableAntigen SynonymsGap junction gamma 1 protein|GJC1|CX 31.9|Gap junction alpha 7 protein|Connexin-45|Connexin 45|Gap junction gamma-1 protein|Gap junction protein alpha 7|DKFZp686P0738|GJD3|CXG1_HUMAN|Connexin 45 / GJA7|gamma 1|Gap junction protein|GJA7|Gap junction protein alpha 7 45kDa|CX 45|Connexin45|Gap junction alpha-7 protein|Cx45|GJA 7|CX31.9|45kDaClonalityPolyclonalConjugationUnconjugatedReactivityHuman, Rat, MouseHostRabbitGene IDP36383IsotypeIgGWestern BlotYesELISAYesImmunogenSynthetic peptide of Human GJC1FormLiquidPurificationAntigen affinity purifiedStorage BufferPBS, 0.05% sodium azide, 40% glycerol, pH 7.4.Storage TemperatureUpon receipt, store at –20 or –80 °C. Avoid repeated freeze.Shipping TemperatureShipped on blue iceNew Product